Ace-Burlitch Inert Atmosphere System, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with Ace no-air labware or suitably adapted conventional glassware
Complete instructions on assembly, leak testing and preparation of the system are included. Components can be ordered separately.
Customers Who Bought This Also Bought