Variable Bore Cap Adapters, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Adapters for replacing rubber seals in variable-bore Omnifit fittings.
- Chemically inert
- Zero dead volume
- PTFE space-filling cone included
Supplied with PTFE space-filling cone. Chemically inert and zero dead volume devices. To fit an Omnifit adapter, the standard cap from any smaller size Omnifit fitting is removed with the O-rings and the adapter screwed onto the fitting in place of the cap.
Customers Who Bought This Also Bought