Solid State Motor Controller, Ace Glass Incorporated
# de catalogue 89077-716
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Solid State Motor Controller
- Cat. no.:89077-716
- Supplier No.:13530-10
Spécifications
A propos de cet article
A compact, lightweight, solid state, ring stand mountable control unit, both AC and DC outlets.
AC output: 120 volts, 10 amps. DC motors give better all- around performance than most other solid state controllers because rectification is balanced full-wave instead of half-wave power. DC output: 0 to 120 volts, 6 amps. Features top Fwd-Off-Rev. motor switch. Rated 1 hp for use with most stirrer motors. Not for use with shunt wound DC motors.
Pulsed AC gives better low speed torque than voltage dropping devices such as rheostat or transformer.
Input voltage: 115 VAC, 60 Hz.
Customers Who Bought This Also Bought