White Silicone Septum with PTFE Bonded Face, Ace Glass
# de catalogue 89077-550
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Ø:32 mm
- Description:Septum
- Thickness:2 mm
- Cat. no.:89077-550
- Supplier No.:12913-40
Spécifications
A propos de cet article
Fits GPI 33-430 phenolic caps
PTFE faced silicone resists coring when punctured via syringe needle. For use with the Screw Caps, Black Plastic, by Ace Glass.
Customers Who Bought This Also Bought