iMiniPlus® Dry Ice Loggers, Cryopak Verification Technologies
Fournisseur: Cryopak Verification Technologies
Certificats
A propos de cet article
The iMiniPlus® Dry Ice logger can be programmed to measure extreme temperatures within the range of -100°C to 40°C (-148°F to 104°F)
- Start and stop buttons on the face of the unitx
- Bookmarking capabilities
- Large LCD display with multifunctional viewing
- Large memory capacity up to 8040 readings
- Polycarbonate housing
Devices have a 1 m cable that connects the stainless steel temperature probe to the data logger. Available as a single-trip or multi-use unit.
It is the ideal solution for ensuring safe transport and storage of perishables held in dry ice.
The data logger box must not be inserted inside the extreme cold environment for risk of stopping the LCD screen from functioning, but particularly of severely damaging the iMiniPlus Dry Ice battery. The probe must therefore be inserted in the environment to be monitored and the box left outside it.
Certifications: ISO9001:2008, CE, RoHS, EN12830.
Spécifications
- Catalog No:
- 97017-230
- 10033-818
- Alarm:
- High and low alarm configuration, 2 thresholds
- High and low alarm configuration, 2 thresholds
- Protection class:
- IP 65
- IP 65
- Battery life:
- 1 - 2 years
- 1 - 2 years
- W×D×H:
- 83×57×18 mm
- 83×57×18 mm
- Data memory - No. of readings:
- 8048
- 8048
- Batteries:
- User resplaceable, lithium button cell (CR 2450), 3 V
- User resplaceable, lithium button cell (CR 2450), 3 V
- Start delay:
- Yes
- Yes