Epoxy Adhesive, Ace Glass Incorporated
# de catalogue 89059-444
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Epoxy adhesive, 30 ml
- Cat. no.:89059-444
- Supplier No.:7560-10
Spécifications
A propos de cet article
A two component epoxy system, which cures to a chemical resistant film.
- Cures to a chemical resistant film
- Bonds tightly
- 30mL jar
The epoxy is used to tightly bond a PTFE sleeve to a glass joint. Supplied in 30 mL jars with the activator in a separate container. The 30 mL supply will cement at least 25 joints depending on size.
Customers Who Bought This Also Bought