Imhoff Cone, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Imhoff sedimentation cone for the determination of settleable matter in water
Either blunt tip or Ace-Thred tip for environmental applications. Graduated from 0 - 2 mL in 0.1mL divisions, 2 - 10 mL in 0.5 mL divisions, 10 - 40 mL in 1 mL divisions, 40 - 100 mL in 2 mL divisions
Customers Who Bought This Also Bought