iMini® Temperature Logger, Cryopak Verification Technologies
À propos de cet article
The iMini® series, provided by Cryopak Verification Technologies, is available as a single trip or multi-use data logger
- 24 months warrenty, excluding battery (Multi use loggers), One Trip, excluding battery (Single trip loggers)
- Password protected, (programmable using ConsolePlus Software) encrypted binary file is generated along with PDF
- The multifunctional LCD screen then provides several levels of information:
- The highest temperature, the lowest temperature and the average temperature recorded
- Any alarms triggered
- The time spent outside alarm ranges
- An icon if the battery is depleted
The iMini® Temperature Logger series was developed to encompass a full spectrum of uses, from food processing and warehousing, to blood storage and transport, to biotech clinical trials and beyond. The device can be programmed in either Celsius or Fahrenheit, and downloaded using a USB nonproprietary interface cable.
All Temperature Loggers offer an IP rating of IP 65.
Standard features include a four alarm threshold with LED indicators, large LCD visibility, and start/stop buttons. Enhanced options include NIST traceable calibration and bookmarking capability. This series offers full versatility of memory capacity and sensor array. With five memory options from 8000 to 128,000 readings and four sensor configurations, these temperature recorders adapt to a multitude of industry specific applications.
6 Options Available Below
Product Details & Documents
The iMini® series, provided by Cryopak Verification Technologies, is available as a single trip or multi-use data logger
- 24 months warrenty, excluding battery (Multi use loggers), One Trip, excluding battery (Single trip loggers)
- Password protected, (programmable using ConsolePlus Software) encrypted binary file is generated along with PDF
- The multifunctional LCD screen then provides several levels of information:
- The highest temperature, the lowest temperature and the average temperature recorded
- Any alarms triggered
- The time spent outside alarm ranges
- An icon if the battery is depleted
The iMini® Temperature Logger series was developed to encompass a full spectrum of uses, from food processing and warehousing, to blood storage and transport, to biotech clinical trials and beyond. The device can be programmed in either Celsius or Fahrenheit, and downloaded using a USB nonproprietary interface cable.
All Temperature Loggers offer an IP rating of IP 65.
Standard features include a four alarm threshold with LED indicators, large LCD visibility, and start/stop buttons. Enhanced options include NIST traceable calibration and bookmarking capability. This series offers full versatility of memory capacity and sensor array. With five memory options from 8000 to 128,000 readings and four sensor configurations, these temperature recorders adapt to a multitude of industry specific applications.
Specifications for Product Family
Spécifications
- Catalog No:
- 10033-794
- 10033-796
- 10033-804
- 10033-798
- 10033-800
- 10033-802
- Sensor:
- Int. sensor
- Int. sensor
- Int. sensor (digital temperature, humidity sensor)
- Ext. sensor
- Int. + ext. sensor
- 2 ext. sensors
- Alarm:
- 4 thresholds; 2 red LEDs, 2 blue LEDs
- 4 thresholds; 2 red LEDs, 2 blue LEDs
- 4 thresholds; 2 red LEDs, 2 blue LEDs
- 4 thresholds; 2 red LEDs, 2 blue LEDs
- 4 thresholds; 2 red LEDs, 2 blue LEDs
- 4 thresholds; 2 red LEDs, 2 blue LEDs
- Interface:
- USB
- USB
- USB
- USB
- USB
- USB
- Software:
- Console Plus
- Console Plus
- Console Plus
- Console Plus
- Console Plus
- Console Plus
- Protection class:
- IP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINI - IP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINI - IP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINI - IP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINI - IP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINI - IP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINI
- Battery life:
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- Response time:
- T90 of 20 min in moving air
- T90 of 20 min in moving air
- T90 of 20 min in moving air
- T90 of 5 min in moving air
- T90 of 20 min in moving air (Int. sensor) T90 of 5 min in moving air (Ext. sensor)
- T90 of 5 min in moving air
- Weight:
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- W×D×H:
- 83×57×18 mm
- 83×57×18 mm
- 83×57×18 mm
- 83×57×18 mm
- 83×57×18 mm
- 83×57×18 mm
- Data memory - No. of readings:
- 8000
- 8000
- 16000
- 8000
- 8000
- 8000
- Batteries:
- 3.0 V, user replaceable (Panasonic only)
- 3.0 V, user replaceable (Panasonic only)
- 3.0 V, user replaceable (Panasonic only)
- 3.0 V, user replaceable (Panasonic only)
- 3.0 V, user replaceable (Panasonic only)
- 3.0 V, user replaceable (Panasonic only)
- Start delay:
- 1 min. - 99 days
- 1 min. - 99 days
- 1 min. - 99 days
- 1 min. - 99 days
- 1 min. - 99 days
- 1 min. - 99 days
Spécifications
Product Family Options
Informations sur le produit
- DésignationRangeRésolutionAccuracyAvailabilityPrice
- iMini® Temperature Logger, Single TripT: –40...80 °C (NTC)T: 0.1 °C (°F)T: 0.5 °C (–40...–10 °C), 0.3 °C (–10...80 °C), ±3% (0 - 100%)Log in pour la disponibilitéEach$132.01Supplier Number: MX-ST-S-8-LSpécifications:
More
Specifications
Cat. No.10033-794SensorInt. sensorAlarm4 thresholds; 2 red LEDs, 2 blue LEDsInterfaceUSBSoftwareConsole PlusProtection classIP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINIBattery life1 - 2 yearsResponse timeT90 of 20 min in moving airWeight70 gW×D×H83×57×18 mmData memory - No. of readings8000Batteries3.0 V, user replaceable (Panasonic only)Start delay1 min. - 99 days - iMini® Temperature Logger, Multi TripT: –40...80 °C (NTC)T: 0.1 °C (°F)T: 0.5 °C (–40...–10 °C), 0.3 °C (–10...80 °C), ±3% (0 - 100%)Log in pour la disponibilitéEach$217.20Supplier Number: MX-IN-S-8-LSpécifications:
More
Specifications
Cat. No.10033-796SensorInt. sensorAlarm4 thresholds; 2 red LEDs, 2 blue LEDsInterfaceUSBSoftwareConsole PlusProtection classIP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINIBattery life1 - 2 yearsResponse timeT90 of 20 min in moving airWeight70 gW×D×H83×57×18 mmData memory - No. of readings8000Batteries3.0 V, user replaceable (Panasonic only)Start delay1 min. - 99 days - iMini® Humidity and Temperature Logger, Multi TripT: –40...80 °C (NTC)
RH: 0 - 100%T: 0.1 °C (°F)T: 0.5 °C (–40...–10 °C), 0.3 °C (–10...80 °C), ±3% (0 - 100%)
RH: ±3% (10 - 90%), ±4% (˂10% and ˃ 90%)Log in pour la disponibilitéEach$457.71Supplier Number: MX-HS-S-16-LSpécifications:More
Specifications
Cat. No.10033-804SensorInt. sensor (digital temperature, humidity sensor)Alarm4 thresholds; 2 red LEDs, 2 blue LEDsInterfaceUSBSoftwareConsole PlusProtection classIP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINIBattery life1 - 2 yearsResponse timeT90 of 20 min in moving airWeight70 gW×D×H83×57×18 mmData memory - No. of readings16000Batteries3.0 V, user replaceable (Panasonic only)Start delay1 min. - 99 days - iMini® Temperature Logger, Multi TripT: –40...80 °C (NTC)T: 0.1 °C (°F)T: 0.5 °C (–40...–10 °C), 0.3 °C (–10...80 °C), ±3% (0 - 100%)Log in pour la disponibilitéEach$291.64Supplier Number: MX-OE-S-8-LSpécifications:
More
Specifications
Cat. No.10033-798SensorExt. sensorAlarm4 thresholds; 2 red LEDs, 2 blue LEDsInterfaceUSBSoftwareConsole PlusProtection classIP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINIBattery life1 - 2 yearsResponse timeT90 of 5 min in moving airWeight70 gW×D×H83×57×18 mmData memory - No. of readings8000Batteries3.0 V, user replaceable (Panasonic only)Start delay1 min. - 99 days - iMini® Temperature Logger, Multi TripT: –40...80 °C (NTC)T: 0.1 °C (°F)T: 0.5 °C (–40...–10 °C), 0.3 °C (–10...80 °C)Log in pour la disponibilitéEach$326.68Supplier Number: MX-1E-S-8-LSpécifications:
More
Specifications
Cat. No.10033-800SensorInt. + ext. sensorAlarm4 thresholds; 2 red LEDs, 2 blue LEDsInterfaceUSBSoftwareConsole PlusProtection classIP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINIBattery life1 - 2 yearsResponse timeT90 of 20 min in moving air (Int. sensor) T90 of 5 min in moving air (Ext. sensor)Weight70 gW×D×H83×57×18 mmData memory - No. of readings8000Batteries3.0 V, user replaceable (Panasonic only)Start delay1 min. - 99 days - iMini® Temperature Logger, Multi TripT: –40...80 °C (NTC)T: 0.1 °C (°F)T: 0.5 °C (–40...–10 °C), 0.3 °C (–10...80 °C), ±3% (0 - 100%)Log in pour la disponibilitéEach$393.53Supplier Number: MX-2E-S-8-LSpécifications:
More
Specifications
Cat. No.10033-802Sensor2 ext. sensorsAlarm4 thresholds; 2 red LEDs, 2 blue LEDsInterfaceUSBSoftwareConsole PlusProtection classIP 65 (with plastic cap over USB connector) for temperature iMINI
IP 51 for humidity iMINIBattery life1 - 2 yearsResponse timeT90 of 5 min in moving airWeight70 gW×D×H83×57×18 mmData memory - No. of readings8000Batteries3.0 V, user replaceable (Panasonic only)Start delay1 min. - 99 days
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- NIST Traceable Temperature Certification - 2 Pts.NIST Traceable Temperature Certification - 2 Pts.10033-954Log in pour la disponibilitéEach - $312.22
Informations sur le produit
- Catalog #AvailabilityPrice
- USB Cable
Informations sur le produit
- Catalog #AvailabilityPrice
- Console Plus Software
Informations sur le produit
- Catalog #AvailabilityPrice
- NIST Traceable Humidity Certification - 2 pts