Flush Seal Bottom Drain Valve Assembly with Pin, Ace Glass Incorporated
# de catalogue 71001-526
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:PTFE Valve Assembly, Flush Seal with Top Pin, Complete Valve Assembly 1" Beaded Pipe
- Cat. no.:71001-526
- Supplier No.:6482-20
Spécifications
A propos de cet article
Redesigned flush-seal type valve with the top of the valve having a 1-1/2" x 3/8" O.D
pin that extends upward. The pin fits into the receptacle on the bottom of certain ACE agitators. This assembly is designed to reduce shaft wobble or “flexing” on the larger reactors. Top opening is 2-inch beaded pipe and the side drain port is 1-inch beaded pipe. Uses clamp from product ID#89064-666.
Customers Who Bought This Also Bought