Photochemical Cell with Removable Window, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Straight, borosilicate glass, photochemical cell with threaded end fittings for use with cell window holder
The ease in removing these holders makes cleaning the cell easier. Available with single or plain port or with single, straight, 2 mm bore, glass stopcock. Not supplied with windows or holders. Please state overall length dimension, otherwise we will supply an overall length of 12". Port will be centered between threaded ends unless requested otherwise.
Customers Who Bought This Also Bought