Photochemical Quartz Immersion Well without Joint, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Double-walled, quartz, immersion well for use with reactors and lamps only
The well offers a greaseless connection and can be adjusted to desired height. Comes with or without #7 Ace Thred bushing. The Ace-Thred means easier dismantling since there is no chance of freezing.
A small diameter inner tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well is used with 89059-230 nylon bushing with FETFE O-Ring to form a compression type seal with threaded reaction vessels.
Customers Who Bought This Also Bought