Photochemical Jacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Jacketed reaction vessel designed to enable cooling of reactant material during photolysis
Constructed of borosilicate glass to accommodate the immersion wells. With one standard taper 14/20 angled joint for sparger tube, one standard taper 24/40 vertical joint for condenser and one #7-Ace-Thred side arm for thermometer. Center joint is #50 Ace-Thred. Bottom of reactor is flat to allow use of stir bar. Volumes indicated are total volumes. Volume in reactive area of lamp is approximately 40-50% of total volume. Size C hose connections. Listing is vessel only.
Customers Who Bought This Also Bought