Photochemical Unjacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Reaction vessel fabricated of borosilicate glass to accommodate the immersion wells only, With #50 Ace-Thred at top which accepts a 7506-14 bushing and FETFE O-Ring to form a compression seal with the immersion well, one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one #7-Ace-Thred side arm for thermometer
Bottom of reactor is flat to allow use of magnetic stirrer. Volumes indicated are total volumes. Working volume in reactive area of lamp is approximately 40-50% of total volume. Complete unit consists of vessel, sparger tube, PTFE stir bar, threaded nylon thermometer bushing, #50 nylon bushing and three meters of 4.8 mm (3/16") I.D. PTFE tubing.
Ordering information: Complete Units Include:
Vessel
89060-570 Sparger Tube
89077-842 PTFE Stir Bar, 38mm X 8mm
89051-122 #7 Nylon Thermometer Bushing
89059-230 #50 Nylon Bushing
PTFE Tubing, 4.8 mm I.D. x 3 m
Vessel
89060-570 Sparger Tube
89077-842 PTFE Stir Bar, 38mm X 8mm
89051-122 #7 Nylon Thermometer Bushing
89059-230 #50 Nylon Bushing
PTFE Tubing, 4.8 mm I.D. x 3 m
Customers Who Bought This Also Bought