Kimble® Disposable Pasteur Pipettes, Soda Lime, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These pipettes feature straight-cut tips and smooth, fire-polished tops to facilitate the attachment of bulbs.
- Made of soda-lime glass
- Available with cotton-plugged or unplugged
They are excellent for use in blood banks, hematology, chemistry, bacteriology and other general lab applications.
Frequently Bought Together