Plant Growth Chambers, Thermo Scientific
# de catalogue 27475-053
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Growth Chamber, Countertop Model
- Cat. no.:27475-053
- Electrical:120 V, 60 Hz
- Capacity:396 L
- Supplier No.:846
Spécifications
A propos de cet article
Grow plants in the classroom with the Thermo Scientific™ Plant Growth Chamber, designed for botanical and agricultural research applications including seed germination studies, plant pathology, plant tissue culture studies and plant genetic research
Chamber accommodates plant, animal and culture studies in the classroom.
Customers Who Bought This Also Bought