ContactUsLinkComponent
Dual Motor Speed and Power Controller, Solid State, Ace Glass Incorporated
Dual Motor Speed and Power Controller, Solid State, Ace Glass Incorporated
# de catalogue 89077-718
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
13532 DUAL MOTOR SPEED AND POWER CONTROLLER, Solid State
Dual Motor Speed and Power Controller, Solid State, Ace Glass Incorporated
# de catalogue 89077-718
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Input: 115VAC, 60Hz
    Output: 0-120VAC, 10A and 0-130VCD, 3A
  • Cat. no.:
    89077-718
  • Supplier No.:
    13532-10
Spécifications
A propos de cet article
Comes with a buffered load control and DC filtration to provide more torque at higher speeds and a higher top speed than motor rating
A compact 8.9 x 8.9 x 17.8 cm (31/2 x 31/2 x 7"), lightweight 1 Kg. (21/4 lbs.) solid state control with rugged control regulation and rectification circuitry. Two output sockets AC and DC which work in conjunction. Fwd-Off-Rev. DC control switch only. Fused for AC and DC outputs, 10 amps and three (3) amps respectively. Rear ring stand clamp for easy mounting and access. Light blue modern type case with protective finish. Red pilot light. Large dial plate with 0-100 divisions. Red control knob with Click-Off. Heavy duty 1.8 meter neoprene, three-wire power cord with NEMA plug.
Ratings: AC three-prong output socket, standard NEMA type, 1200 watts. 0-120 volts, variable, 60 Hz. maximum 10 amps. DC four-prong output socket, “cinch” type. 360 watts, 0-150 volts, variable, filtered approximately ½ hp. max. 3 amps.
Customers Who Bought This Also Bought