Volumetric Bulbs, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Fills and pours automatically, rapidly.
- One head for all capacities, all volumes
- Short tipping angle, interchangeable volumetric bulbs give reproducible volumes
Bulb joint is a standard taper 14/20, head and flask standard taper 29/42.
Complete apparatus consists of head, flask, volumetric bulb, clip, two 19 mm (3/4") springs, one 38 mm (1½") spring.
Customers Who Bought This Also Bought