Nalgene® Biotainer® Polycarbonate Bottles and Vials, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing, and associated validations.
- Sterile
- Ready-to-use
- Excellent for freezing applications
Nalgene® InVitro™ Biotainer® bottles and vials are designed specifically for pharmaceutical and biological manufacturers.
Customers Who Bought This Also Bought