LabMarkers, Micronova
Supplier: Micronova
Certificates
About this item
LabMarkers feature permanent, quick drying, alcohol-resistant ink that marks on cold or wet surfaces, plastic bags, or disposable labware.
- Fine tip
- Black ink
- Fast drying
- Permanent on most surfaces
- Available gamma irradiated
Marks will not smear on reusable glass, metal or porcelain labware but can be scrubbed off. This fine tipped marker's ink is permanent on most surfaces.
Customers Who Bought This Also Bought