Storage Tube, Micro Schlenk, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This tube is used for storage or simple reactions.
- Stopcock on side arm is 1 mm bore glass
- Item supplied with holed cap, O-ring and septa for sealing or syringe retrieval
With a standard taper 10/10 or standard taper 14/10 outer joint externally threaded for making an O-ring-cap-seal connection with mating inner joint.
Customers Who Bought This Also Bought