Glass Beakers, Round, Phipps & Bird
# de catalogue 89370-880
Fournisseur: Phipps & Bird
Small Business Enterprise
Spécifications
- Capacity:1000 ml (34 oz.)
- Cat. no.:89370-880
- Supplier No.:7790-620
Spécifications
A propos de cet article
Round glass beakers for water treatment.
Customers Who Bought This Also Bought