Micro/Mini-Lab Distillate Take-Off Adapter, Ace Glass Incorporated
# de catalogue 89075-146
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Cat. no.:89075-146
- Top ST Joint Size:[ST] 14/10
- Supplier No.:9563-07
Spécifications
A propos de cet article
The top joint of this adapter is a combination cap, thread and a standard taper joint to accommodate and seal other microscale apparatus.
- O.D. of top joint has an external thread
- I.D. at top is a ground [ST] outer joint
- 105° angle
- Cap and o-ring included
The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground standard taper 14/10 outer joint. 105° angle. Supplied with cap and o-ring.
Customers Who Bought This Also Bought