Borosilicate Glass Standard Taper Inner Ground Joint with O-Ring Groove, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Adapter features a standard taper inner member ground joint with a groove for an O-Ring.
- Leak-proof at high vacuum
- O-ring permits use without grease
- One FETFE o-ring included
For use with outer members. Adapter is leak-proof at high vacuum (up to 1 x 10-8 mm Hg.). O-Ring permits use without grease. Supplied with one FETFE o-ring.
Customers Who Bought This Also Bought