Axygen® Prebarcoded PCR Plates, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Axygen® prebarcoded PCR plates are manufactured to stringent specifications under strict process controls.
Plates are manufactured from 99.9% pure virgin polypropylene. Barcode is located on the long side of the plate between well H1-H12. Plates are certified RNase-/DNase-free, and nonpyrogenic. 96-Well and 384-Well options are available.
They are assembled via automation to ensure consistent performance and quality. Ultra-thin and consistent wall thickness allows precise thermal transfer.
Spécifications
- Catalog No:
- 10032-810
- 10032-812
- 10032-808
- 10032-804
- 10032-806
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa