Epoxy Coated Pressure Reactor Manifold, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Pressure manifold provides for safer operation of Ace pressure filter reactors.
Manifold features an epoxy coating, 0-60 psi pressure gauge with stainless steel internals, 0-50 psi adjustable relief valve and over-pressure safety rupture disc. Ace-Thred to NPT connections are used throughout.
Customers Who Bought This Also Bought