SP Bel-Art Swing Jaw™ Tubing Clamps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Zinc-plated steel tube clamp provides a non-slip closure for all flexible tubing – even heavy-walled vacuum lines.
- Zinc-plated steel tube clamp construction
- Can be installed without disconnecting tubing
- Smooth clamping surfaces
A knurled nut compresses against a swinging jaw with smooth clamping surfaces. Can be installed and removed without disconnecting tubing.
Customers Who Bought This Also Bought