Lock-N-Seal™ Water Sampling Vials, Excel Scientific
Fournisseur: Excel Scientific
Certificats
A propos de cet article
Lock-N-Seal Vials with ties are for water sampling.
- Vials are available with or without Sodium Thiosulfate pill
They feature a 100 ml EPA fill line for volume estimation.
Customers Who Bought This Also Bought