N.I.S.T Factory Certificate of Accuracy, Ace Glass Incorporated
# de catalogue 89051-908
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Certificate
- Cat. no.:89051-908
- Supplier No.:5697-10
Spécifications
A propos de cet article
This certificate gives corrections at the test points and also certifies that the delivery time and other physical requirements are within the specifications set by N.I.S.T
The method used in determining the corrections is recommended by the National Bureau of Standards. Suitable for all Ace or Corning burets, tubes, flasks or measuring vessels. All capacities priced the same
Customers Who Bought This Also Bought