Bench Scale Jacketed Filter Reactor, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Jacketed filter reactor systems include all of the necessary components to perform single- or multi-step reactions and filtrations in the same vessel
Pressure version maximum pressure is 35psig at 100°C (212°F). Maximum jacket pressure is 8 psig.
Pressure version; maximum pressure is 35psig at 100°C. Maximum jacket pressure is 8psig.
Systems with Ace-Thred heads are pressure rated to 35psi at 100°C. System sizes vary from 150mL to 6000mL with 60mm (2.4"), 100mm (4"), or 150mm (6") Duran® flanged heads. Reactor bodies feature ball joint inlets/outlets or Ace-Threds, which can be ordered separately. Stand, shelf, chain clamp, and clamp holder not included.
Customers Who Bought This Also Bought