SP Bel-Art Heavy Duty Faucets, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
A needle-type, Teflon®O-Ring seal inside this polyethylene faucet prevents back pressure liquid from escaping through the operating knob.
- Polyethylene construction
- Needle-type seal
- Secure against back pressure liquid escaping
The outlet accepts 16 mm I.D. (5/8") tubing.
Faucet 47750-016 fits standard steel drums. Faucet 47750-018 has a male threaded outlet that accommodates garden hose coupling standard 19 mm – 4.53 threads per cm (3/4" – 11.5 threads per inch).
Customers Who Bought This Also Bought