SP Bel-Art Tubing Sinkers, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Teflon® sinkers keep tubing at the bottom of a container.
- Hose barb ends
- Solvent-resistant
- Autoclavable
Ideal for use with pumps and dispensers, these sinkers have hose barb ends for simple tubing connection.
Customers Who Bought This Also Bought