CurTec Polypropylene Lids for Packo Containers, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Packo Lids are available with an integrated gasket, or plain for use with inner seals also (sold separately). This High Performance Packaging line provides the highest level of product protection and added value through quality, functionality, ease of use, and design.
- Quarter-turn screw lid
- Water vapor tight
- Integrated tamper evident strip
- UN certified with Packo jars
Suggested for use with pharmaceuticals, fine chemicals, food ingredients, and hazardous waste. Airtight design prevents moisture contamination.
Customers Who Bought This Also Bought