SP Bel-Art Evaporating Dish, Bel-Art Products, a part of SP
Catalog # 47748-186
Supplier: Bel-Art Products, a Part of SP
Specifications
- Depth:4.2 cm
- Ø:10 cm
- Capacity:175 ml
- Cat. no.:47748-186
- Supplier No.:409250000
Specifications
About this item
Smooth polypropylene dish with easy pour spout holds up to 175 mL.
- Polypropylene construction
- Reusable
- Autoclavable
Useful for titration and evaporation, the dish resists acids and alkalis and is autoclavable at 121°C (250°F). It can also be used as a reusable weighing dish.
Customers Who Bought This Also Bought