SP Bel-Art Leveling Table, Bel-Art Products, a part of SP
# de catalogue 47750-926
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Dimensions:20.3 x 20.3 x 0.95 cm (8 x 8 x ⅜")
- Description:Leveling Table
- Cat. no.:47750-926
- Supplier No.:183100000
Spécifications
A propos de cet article
This clear and rigid acrylic level surface helps in pouring immunological plates, horizontal agarose gels, and acrylamide sequencing gels.
- Clear, rigid acrylic construction
- Adjustable legs
- Inset leveling bulb assists in accurate adjustment
Each leg is adjustable, and an inset leveling bulb below the working surface assists in accurate adjustment.
Customers Who Bought This Also Bought