SP Bel-Art Wash Bottles, Red, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
The bright red color of these wash bottles helps identify hazardous contents.
- LDPE bottle construction
- Polypropylene closure included
- Bright red color identifies hazardous contents
Bottles are manufactured from low-density polyethylene and are supplied with a polypropylene closure.
Customers Who Bought This Also Bought