Heating Mantle, Pear Shaped, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Heating Mantle for use with pear-shaped flasks
Mantle withstands 400 Degrees Celsius internal operating temperatures. Supplied with 4 foot 2 wire cord with locking connector. Most mantles are less than 115 volts and must be operated through a power control. Use with the following power controllers,Ace Glasses' Solid State Voltage Controller, Powerstat Variable Transformer, or POWERSTAT. Not available in 230V.
Customers Who Bought This Also Bought