Aviation Freeze Point Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
For determining the freezing point of separated solids in aviation fuels
Thermometer not supplied. The Complete Kit includes one rubber stopper, jacketed sample tube, brass packing gland, collar with side tube, collar without side tube, stirring rod, and non-silvered vacuum flask.
Customers Who Bought This Also Bought