Padded Process Pipe Pipe Hanger Without Foot, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
All hangers are zinc coated and have factory applied plastic sleeve to prevent glass-to-metal contact
Use one hanger every 8 or 10 feet. Support all valves and gauges individually.
Frequently Bought Together