Color Stability Tube, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Used in tentative AOCS Method L-15A-58 for measuring the processing color stability of fatty acids.
Customers Who Bought This Also Bought
Used in tentative AOCS Method L-15A-58 for measuring the processing color stability of fatty acids.