Interchangeable Tissue Grinder, Ace Glass
Supplier: Ace Glass
Certificates
About this item
This is a precision ground instrument for macerating tissue
Rod and barrel are interchangeable. Rod is ground and its entire length and can be reversed for double life. Barrel clearance of .13mm to .2mm meets G.S.A. specifications
Customers Who Bought This Also Bought