Beaded Process Pipe, Mitered 90° Elbow, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Beaded Process Pipe with mitered 90° elbow is made of borosilicate glass
Pipe is durable in all acids except hydrofluoric or phosphoric acids with fluorides. Some alkalies attack above 120°F and above a pH of 11.
Operating temperature: 230 °C
Spécifications
- Catalog No:
- 89064-504
- 89064-506
- 89064-502
- 89064-498
- 89064-500
- Model:
- 89064-504
- 89064-506
- 89064-502
- 89064-498
- 89064-500
- Working pressure:
- 50 PSI
- 30 PSI
- 75 PSI
- 75 PSI
- 75 PSI