Micro Heat Transfer Block, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Aluminum heat transfer block for use on top of stirrer-hot plate to effect better heat transfer
Block has three bowl-shaped recesses for 3 and 5 mL thin-walled vials and 5, 10 and 25 mL round-bottom flasks; one 20 mm straight-thru hole, one deep bowl-shaped hole for 10 mm test tubes and one hole for a thermometer. Measures 3 1/2 x 3/4".
Customers Who Bought This Also Bought