BEST® Nitrile Disposable Gloves, Showa
Fournisseur: Showa
Certificats
A propos de cet article
BEST® gloves are for manufacturing, light-duty maintenance and cleanup, intricate parts handling, laboratory analysis/technical work, automotive applications.
- Gloves feature a rolled cuff and a textured grip
- Economy grade
Spécifications
- Catalog No:
- 10809-738
- 12000-152
- 10119-994
- 12000-150
- 12000-148
- 12000-154
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Finish:
- Textured
- Textured
- Textured
- Textured
- Textured
- Textured