Temperature Monitoring Systems, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This monitoring system is a complete package to enable the user to monitor the temperature of jacketed 10, 30, and 50 L cylindrical Pilot Plant Reactor
Complete with LED display and recorder output, PTFE covered Type J Temperature Sensor with detachable lead, PTFE Adapter, adapters to secure temperature sensor in head of reactor, and Nylon Bushing. Thermocouple only can be ordered.
Ordering information: The Complete Temperature Monitoring System for 10 or 20 mL Cylindrical comes with Thermometer, #7 Adapter, "J" Thermocouple Sensor, Sensor Cord. The Complete Temperature Monitoring System for 30 L comes with Thermometer, Glass Adapter, PTFE Adapater, Nylon Bushing, "J" Thermocouple Sensor, Sensor Cord.
Customers Who Bought This Also Bought