Beaded Process Pipe, U Bend, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Beaded process pipe U bend.
- For all except hydrofluoric or phosphoric with fluorides durability acids
- Some durability alkalies attack above 120 °F and above pH11
Shock difference: 160 °C
Thermal expansion: 32 x 10-7 °C
Conductivity: 0.73 BTU/ hr. ft.2/F°/ft. at 212 °F
Operating temperature: Freezing point to 450 °C
Spécifications
- Catalog No:
- 89064-656
- 89064-648
- 89064-650
- 89064-652
- 89064-654
- 89064-644
- 89064-646
- Model:
- 89064-656
- 89064-648
- 89064-650
- 89064-652
- 89064-654
- 89064-644
- 89064-646
- Working pressure:
- 50 PSI
- 100 PSI
- 75 PSI
- 75 PSI
- 75 PSI
- 100 PSI
- 100 PSI