SP Bel-Art HPLC Reservoir Secondary Containers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Safeguard expensive HPLC solvent reservoirs against damage or breakage while in use or during storage with these durable polyethylene HPLC Reservoir Secondary Containers.
- Durable polyethylene construction
- Chemically resistant
- Available in a variety of sizes
These containers are resistant to most chemicals and are available in 5 l, 10 l and 20 l sizes to accommodate popular HPLC solvent reservoirs.
In addition to protecting the HPLC reservoirs, the containers serve as secondary containment to prevent solvent spills in the work area. The large, unobstructed opening allows easy viewing of the HPLC reservoir, and the round shape of the containers minimizes wasted space on the benchtop.
Frequently Bought Together