SP Bel-Art LabLens® Liquid, Bel-Art Products, a part of SP
Catalog # 13197-193
Supplier: Bel-Art Products, a Part of SP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:LabLens® Liquid Cleaning Solution
- Size:473 ml (1 pt.)
- Cat. no.:13197-193
- Supplier No.:248330000
Specifications
About this item
Lens cleaning solution in a 473 ml (1 pt.) polyethylene bottle with spray pump.
- Polyethylene bottle construction
- Spray pump allows easy application
Bottle contains a non-silicone fluid that protects, polishes, anti-stats, and anti-fogs both glass and plastic lenses.
Customers Who Bought This Also Bought