SP Bel-Art Graduated Cylinders, Clear TPX®, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Clear TPX® (PMP) plastic cylinders are non-wetting to prevent a concave meniscus, making the raised numeral, single scale graduations easy to read.
- Superior chemical resistance
- Shatter-proof
- Safe for use to 175 ºC (350 ºF)
- Steam autoclavable at 121 ºC (250 ºF)
- Made of TPX® Brand polymethylpentene for easy viewing
They are shatter-proof with a wide octagonal base to prevent tipping and rolling.
Specifications
- Catalog No:
- 16126-584
- 16126-580
- 16126-586
- 24780-376
- 13197-208
- 30617-660
- 13197-206
- 24780-378
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall