SP Bel-Art Rimless Centrifuge Tubes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Rimless centrifuge tubes are designed for use in centrifuges up to 50000 g without refrigeration.
- Polypropylene construction
- Rimless
- Round-bottomed
- Autoclavable
These polypropylene tubes are rimless, round-bottomed, and autoclavable at 121 °C (250 °F).
Customers Who Bought This Also Bought