SP Bel-Art Microcentrifuge Tube, Bel-Art Products, a part of SP
Catalog # 20901-925
Supplier: Bel-Art Products, a Part of SP
Specifications
- Volume:1.5 ml
- Description:Polypropylene Microcentrifuge Tube, 11 x 42 mm
- Cat. no.:20901-925
- Supplier No.:F19929-0000
Specifications
About this item
These micro sample tubes have a captive plug and will withstand the forces of a microcentrifuge.
- Captive plug design
- Polypropylene construction
- Autoclavable
Polypropylene tubes are steam autoclavable at 121 °C (250 °F).
Frequently Bought Together